missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNH6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116625
This item is not returnable.
View return policy
Description
Positive Samples: 293T, HeLa Immunogen sequence: TGPNTPSSAV SRLAQALLGA EECKVDILYY RKDASSFRCL VDVVPVKNED GAVIMFILNF EDLAQLLAKC SSRSLSQRLL SQSFLGSEGS HGRPGGPGPG TGRGKYRTIS QIPQFTLNFV EFNLEKHRSS STTEIEIIAP HKVVERTQNV T.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. KCNH6 encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit.
Specifications
| KCNH6 | |
| Polyclonal | |
| Unconjugated | |
| KCNH6 | |
| Eag-related gene member 2; eag-related protein 2; Erg2; ERG-2; Ether-a-go-go-related gene potassium channel 2; ether-a-go-go-related protein 2; HERG2; hERG-2; KCNH6; Kv11.2; m-erg2; potassium channel, voltage gated eag related subfamily H, member 6; potassium voltage-gated channel subfamily H member 6; potassium voltage-gated channel, subfamily H (eag-related), member 6; voltage-gated potassium channel subunit Kv11.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 81033 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9H252 | |
| KCNH6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-220 of human KCNH6 (NP_1104061). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction