missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNH6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550775
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total KCNH6 protein.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. KCNH6 encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit.
Specifications
| KCNH6 | |
| Polyclonal | |
| Unconjugated | |
| KCNH6 | |
| Eag-related gene member 2; eag-related protein 2; Erg2; ERG-2; Ether-a-go-go-related gene potassium channel 2; ether-a-go-go-related protein 2; HERG2; hERG-2; KCNH6; Kv11.2; m-erg2; potassium channel, voltage gated eag related subfamily H, member 6; potassium voltage-gated channel subfamily H member 6; potassium voltage-gated channel, subfamily H (eag-related), member 6; voltage-gated potassium channel subunit Kv11.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81033 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9H252 | |
| KCNH6 | |
| Fusion protein corresponding to residues near the N terminal of human potassium voltage-gated channel, subfamily H (eag-related), member 6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction