missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNH2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11578851
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11578851 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11578851 Supplier Invitrogen™ Supplier No. PA3860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA3-860 detects HERG in human samples. PA3-860 has been used successfully in Western blot, immunfluorescence, and immunoprecipitation procedures. By Western blot this antibody detects the ~135 kDa and ~155 kDa forms of HERG. The PA3-860 immunogen corresponds to a cDNA fragment of HERG encoding 181 amino acid residues from the C-terminus.

Human ether-a-go-go related gene (HERG) encodes the pore-forming alpha subunit of the delayed rectifier potassium channel IKr. There are two N-terminal splice variants of HERG include the full-length isoform 1 alpha and the shorter isoform 1 beta. Isoform 1 beta lacks the PAS motif and deactivates at a faster rate than isoform 1alpha. Residues within the C-terminal play a role in channel expression and channel gating, including voltage-dependent activation. HERG is expressed in the heart and is more abundantly expressed in the ventricles than in the atria. Mutations in the gene encoding HERG increase beat-to-beat variability and early after depolarization. These fluctuations facilitate the genesis and propagation of premature heartbeats associated with inheritable long QT syndrome type 2 and short QT syndrome type 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNH2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene KCNH2
Gene Accession No. Q12809
Gene Alias AI326795; cerg; c-ERG; cERG; ether-a-go-go related member 2; delayed rectifier potassium channel; derg; Eag homolog; eag related protein 1; eag-related gene; eag-related protein 1; ERG; ERG1; ERG-1; ether-a-go-go related gene potassium channel 1; ether-a-go-go related protein; ether-a-go-go-related gene potassium channel 1; ether-a-go-go-related potassium channel protein; ether-a-go-go-related protein 1; gp-erg; HERG; H-ERG; HERG1; HERG-1; KCNH2; Kv11.1; LQT; Lqt2; M-erg; Merg1; M-erg1; merg1a; merg1b; potassium channel; potassium channel protein ERG; potassium channel protein ERG long isoform; potassium channel protein ERG short isoform; potassium channel, voltage gated eag related subfamily H, member 2; potassium voltage-gated channel subfamily H member 2; potassium voltage-gated channel, subfamily H (eag-related), member 2; ra-erg; RERG; r-ERG; SQT1; ventricular ERG K+ channel subunit; voltage-gated potassium channel 11.1; voltage-gated potassium channel subunit Kv11.1; voltage-gated potassium channel variant 3; voltage-gated potassium channel, subfamily H, member 2
Gene Symbols KCNH2
Host Species Rabbit
Immunogen Fusion Protein of HERG encoding 181 amino acid residues from the C-terminus.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3757
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.