missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNG1 Polyclonal Antibody
KCNG1 Polyclonal Antibody
Brand: Thermo Scientific PA559033
This item is not returnable.
View return policy
Description
KCNG1 Polyclonal Antibody for IHC
Specifications
| KCNG1 | |
| Polyclonal | |
| Unconjugated | |
| KCNG1 | |
| AW536275; K13; KCNG; KCNG1; kH2; KV6.1; orthologue of H. sapiens potassium voltage-gated channel, subfamily G, member 1 (KCNG1); OTTMUSG00000016048; potassium channel KH2; potassium channel Kv6.1; potassium channel, voltage gated modifier subfamily G, member 1; potassium channel, voltage-gated modifier subfamily G, member 1; potassium voltage-gated channel modifier subfamily G member 1; potassium voltage-gated channel subfamily G member 1; potassium voltage-gated channel, subfamily G, member 1; voltage-gated potassium channel subunit Kv6.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3755 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UIX4 | |
| KCNG1 | |
| Recombinant protein corresponding to Human KCNG1. Recombinant protein control fragment (Product #RP-98323). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction