missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNE4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100586
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KCNE4.
Ancillary protein that assembles as a beta subunit with a voltage-gated potassium channel complex of pore-forming alpha subunits. Modulates the gating kinetics and enhances stability of the channel complex. May associate with KCNQ1/KVLTQ1 and inhibit potassium current.
Specifications
| KCNE4 | |
| Polyclonal | |
| Unconjugated | |
| KCNE4 | |
| AW487937; cardiac voltage-gated potassium channel accessory subunit 4; KCNE4; Minimum potassium ion channel-related peptide 3; minK-related peptide 3; MIRP3; potassium channel subunit beta MiRP3; potassium channel, voltage gated subfamily E regulatory beta subunit 4; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 4; potassium voltage-gated channel subfamily E member 4; potassium voltage-gated channel subfamily E regulatory subunit 4; potassium voltage-gated channel, Isk-related family, member 4; potassium voltage-gated channel, Isk-related subfamily, gene 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23704, 367302, 57814 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WWG9, Q9WTW3 | |
| KCNE4 | |
| A synthesized peptide derived from human KCNE4(Accession Q8WWG9), corresponding to amino acid residues Y87-P137. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction