missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562432
This item is not returnable.
View return policy
Description
Immunogen sequence: STVKSKRREH SNDPYHQYIV EDWQEKYKSQ ILNLEESKAT IHENIGAAGF KMS Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, isk-related subfamily. This member is a small integral membrane subunit that assembles with the KCNH2 gene product, a pore-forming protein, to alter its function. This gene is expressed in heart and muscle and the gene mutations are associated with cardiac arrhythmia.
Specifications
| KCNE2 | |
| Polyclonal | |
| Unconjugated | |
| KCNE2 | |
| 2200002I16Rik; ATFB4; AW048273; cardiac voltage-gated potassium channel accessory subunit 2; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; IL-10R2; Kcne2; LQT5; LQT6; minimum potassium ion channel related peptide 1; minimum potassium ion channel-related peptide 1; MinK-related peptide 1; minK-related peptide-1; MIRP1; Potassium channel subunit beta MiRP1; potassium channel subunit, MiRP1; potassium channel, voltage gated subfamily E regulatory beta subunit 2; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 2; potassium voltage-gated channel subfamily E member 2; potassium voltage-gated channel subfamily E regulatory subunit 2; potassium voltage-gated channel, Isk-related family, member 2; potassium voltage-gated channel, Isk-related subfamily, gene 2; voltage-gated K+ channel subunit MIRP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9992 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6J6 | |
| KCNE2 | |
| Recombinant protein corresponding to Human KCNE2. Recombinant protein control fragment (Product #RP-103104). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction