missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNE2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15784141 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15784141 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15784141 Supplier Invitrogen™ Supplier No. PA556233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MSTLSNFTQT LEDVFRRIFI TYMDNWRQNT TAEQEALQAK VDAE Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, isk-related subfamily. This member is a small integral membrane subunit that assembles with the KCNH2 gene product, a pore-forming protein, to alter its function. This gene is expressed in heart and muscle and the gene mutations are associated with cardiac arrhythmia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNE2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNE2
Gene Accession No. Q9Y6J6
Gene Alias 2200002I16Rik; ATFB4; AW048273; cardiac voltage-gated potassium channel accessory subunit 2; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; IL-10R2; Kcne2; LQT5; LQT6; minimum potassium ion channel related peptide 1; minimum potassium ion channel-related peptide 1; MinK-related peptide 1; minK-related peptide-1; MIRP1; Potassium channel subunit beta MiRP1; potassium channel subunit, MiRP1; potassium channel, voltage gated subfamily E regulatory beta subunit 2; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 2; potassium voltage-gated channel subfamily E member 2; potassium voltage-gated channel subfamily E regulatory subunit 2; potassium voltage-gated channel, Isk-related family, member 2; potassium voltage-gated channel, Isk-related subfamily, gene 2; voltage-gated K+ channel subunit MIRP1
Gene Symbols KCNE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNE2. Recombinant protein control fragment (Product #RP-102558).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.