missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNE1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568274
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Kcne1 is a member of Voltage-gated potassium (Kv) channels that represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of Kv channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The Kcne1 gene encodes a membrane protein which has sequence similarity to the KCNE1 gene product, a member of the potassium channel, voltage-gated, isk-related subfamily. Kcne1, an intronless gene is deleted in AMME contiguous gene syndrome and may be involved in the cardiac and neurologic abnormalities found in the AMME contiguous gene syndrome. Functionally, Kcne1 is a potassium channel ancillary subunit that is essential for generation of some native K(+) currents by virtue of formation of heteromeric ion channel complex with voltage-gated potassium (Kv) channel pore-forming alpha subunits. Kcne1 also functions as an inhibitory beta-subunit of the repolarizing cardiac potassium ion channel KCNQ1. Diseases associated with KCNE1 include Jervell And Lange-Nielsen Syndrome 2 and Long Qt Syndrome 5.
Specifications
| KCNE1L | |
| Polyclonal | |
| Unconjugated | |
| KCNE5 | |
| 1500015C14Rik; AMME syndrome candidate gene 2 protein; AMMECR2; AMMECR2 protein; cardiac voltage-gated potassium channel accessory subunit 5; KCNE1L; KCNE1-like; Kcne5; Mink; MinK-like protein; potassium channel subunit beta MiRP4; potassium channel, voltage gated subfamily E regulatory beta subunit 5; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 5; potassium voltage-gated channel subfamily E member 1-like protein; Potassium voltage-gated channel subfamily E regulatory beta subunit 5; potassium voltage-gated channel subfamily E regulatory subunit 5; potassium voltage-gated channel, Isk-related family, member 1-like; potassium voltage-gated channel, Isk-related family, member 1-like, pseudogene; voltage-gated potassium channel accessory subunit 5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23630, 681190 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UJ90 | |
| KCNE5 | |
| Synthesized peptide derived from human KCNE1L. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction