missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KCNB1 Polyclonal antibody specifically detects KCNB1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | KCNB1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 5 - 10 ug/ml |
| Formulation | PBS |
| Gene Alias | Delayed rectifier potassium channel 1, delayed rectifier potassium channel Kv2.1, DRK1, h-DRK1, h-DRK1 K(+) channel, Kv2.1, potassium channel protein DRK1, potassium voltage-gated channel subfamily B member 1, potassium voltage-gated channel, Shab-related subfamily, member 1, Voltage-gated potassium channel subunit Kv2.1 |
| Host Species | Rabbit |
| Immunogen | Rabbits were immunized with a synthetic peptide derived from the rat Kv2.1 potassium channel conjugated to KLH (Uniprot: Q14721) |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?