missing translation for 'onlineSavingsMsg'
Learn More

KCNB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £513.00

Specifications

Antigen KCNB1
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18681646
View Documents
Novus Biologicals
NBP2-38498-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18170278
View Documents
Novus Biologicals
NBP2-38498
0.1 mL
£513.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KCNB1 Polyclonal specifically detects KCNB1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

KCNB1
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
Q14721
3745
This antibody was developed against a recombinant protein corresponding to amino acids: ECATLLDKAVLSPESSIYTTASAKTPPRSPEKHTAIAFNFEAGVHQYIDADTDDEGQLLYSVDSSPPKSLPGSTSPKFSTGTRSEKNHFESSPLPTSPKFLRQNCIYSTEALTGK
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Signal Transduction
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Delayed rectifier potassium channel 1, delayed rectifier potassium channel Kv2.1, DRK1, h-DRK1, h-DRK1 K(+) channel, Kv2.1, potassium channel protein DRK1, potassium voltage-gated channel subfamily B member 1, potassium voltage-gated channel, Shab-related subfamily, member 1, Voltage-gated potassium channel subunit Kv2.1
KCNB1
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.