Learn More
KCMF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556454
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MSETERQSME SERADRSLFV QELLLSTLVR EESSSSDEDD RGEMADFGAM GCVDIMPLDV ALENLNLKES NKGNEPPPPP L Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
KCMF1 belongs to the KCMF1 family. It contains 1 C2H2-type zinc finger and 1 ZZ-type zinc finger. It has intrinsic E3 ubiquitin ligase activity and promotes ubiquitination.Specifications
KCMF1 | |
Polyclonal | |
Unconjugated | |
KCMF1 | |
1700094M07Rik; clone DEBT-91; DEBT91; debt-91; Differentially expressed in branching tubulogenesis 91; DKFZp434L1021; E3 ubiquitin-protein ligase KCMF1; FGF-induced in gastric cancer; FGF-induced ubiquitin-protein ligase in gastric cancers; FIGC; kcmf1; kcmf1 protein; PCMF; Pmcf; Potassium channel modulatory factor; potassium channel modulatory factor 1; potassium channel modulatory factor DEBT-91; RING-type E3 ubiquitin transferase KCMF1; si:ch211-220b11.7; zgc:64160; zinc finger, ZZ domain containing 1; zKCMF1; ZZ-type zinc finger-containing protein 1; ZZZ1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56888 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P0J7 | |
KCMF1 | |
Recombinant protein corresponding to Human KCMF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only