missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KChIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541075
This item is not returnable.
View return policy
Description
Peptide sequence: QLTDSVDDEF ELSTVCHRPE GLEQLQEQTK FTRKELQVLY RGFKNPGALS Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Voltage-gated potassium (Kv) channels are key components of the potassium currents in the heart and central nervous system. In the heart, Kv4 channels are involved in the repolarization phase of the action potential. In the brain however, these channels prevent reverse-propagation of action potentials. Found associated with Kv4 channels are a group of calcium-binding proteins termed KChIPs (Kv channel interacting proteins). KChIPs are small molecular weight proteins that bind to the cytoplasmic amino termini of Kv4 alpha-subunits and help to modulate its function. There are currently three known KChIPs; KChIP1 expressed in the brain; KChIP2 expressed in the brain, heart, and lung, and has three isoforms, a, b, and c; and KChIP3, also known as calsenilin and DREAM (downstream regulatory elements (DRE)-antagonist modulator) which is expressed in the brain and testis.
Specifications
| KChIP2 | |
| Polyclonal | |
| Unconjugated | |
| KCNIP2 | |
| A-type potassium channel modulatory protein 2; Cardiac voltage-gated potassium channel modulatory subunit; DKFZp566L1246; KCHIP2; KChlP2; Kcnip2; Kv channel interacting protein 2; Kv channel-interacting protein; Kv channel-interacting protein 2; MGC17241; potassium channel auxiliary subunit KCHIP2; potassium channel-interacting protein 2; potassium voltage-gated channel interacting protein 2; si:ch73-173h19.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 30819, 80906 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9JJ69, Q9NS61 | |
| KCNIP2 | |
| Synthetic peptide directed towards the N-terminal of human KCNIP2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction