missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KChIP2 Antibody (OTI3C4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71866CL1
This item is not returnable.
View return policy
Description
KChIP2 Monoclonal antibody specifically detects KChIP2 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| KChIP2 | |
| Monoclonal | |
| CoraFluor 1 | |
| A-type potassium channel modulatory protein 2, cardiac voltage gated potassium channel modulatory subunit, Cardiac voltage-gated potassium channel modulatory subunit, DKFZp566L1246, KChIP2, KCHIP2Kv channel-interacting protein 2, Kv channel interacting protein 2, MGC17241, potassium channel interacting protein 2, Potassium channel-interacting protein 2 | |
| Full length human recombinant protein of human KCNIP2 (NP_775284) produced in E.coli. | |
| 0.1 mL | |
| Vision | |
| 30819 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| OTI3C4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction