Learn More
KCC4/SLC12A7 Antibody (891526), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9030MFV500
This item is not returnable.
View return policy
Description
KCC4/SLC12A7 Monoclonal antibody specifically detects KCC4/SLC12A7 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Monoclonal | |
| 50mM Sodium Borate | |
| 0.1 mL |
| mFluor Violet 500 SE | |
| Mouse | |
| 10723 |