missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCC4 Monoclonal Antibody (891526)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561982
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Mediates electroneutral potassium-chloride cotransport when activated by cell swelling. May mediate K(+) uptake into Deiters' cells in the cochlea and contribute to K(+) recycling in the inner ear. Important for the survival of cochlear outer and inner hair cells and the maintenance of the organ of Corti. May be required for basolateral Cl(-) extrusion in the kidney and contribute to renal acidification. [UniProt].
Specifications
| KCC4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y666 | |
| SLC12A7 | |
| NS0 mouse myeloma cell line transfected with human SLC12A7, Met1-Ser1083. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 891526 | |
| Unconjugated | |
| SLC12A7 | |
| AA408796; D13Ertd261e; electroneutral potassium-chloride cotransporter 4; EMBL:AAH86339.1, ECO:0000312; furosemide-sensitive KCl cotransporter 4; Kcc4; K-Cl cotransporter 4; K-Cl cotransporter KCC4; potassium/chloride transporter KCC4; RGD:1359672}; Slc12a7; slc12a7 {ECO:0000312; solute carrier family 12 (potassium/chloride transporter), member 7; solute carrier family 12 (potassium/chloride transporters), member 7; solute carrier family 12 member 7; solute carrier family 12, member 7 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10723 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction