missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ KCC3/SLC12A6 Antibody, Novus Biologicals™

Product Code. 30047487 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30047487 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30047487 Supplier Novus Biologicals™ Supplier No. NBP329424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCC3/SLC12A6 Polyclonal antibody specifically detects KCC3/SLC12A6 in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCC3/SLC12A6
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Tris-citrate/phosphate buffer, pH 7 to 8
Gene Alias ACCPN, agenesis of corpus callosum and peripheral neuropathy (Andermann syndrome), DKFZp434D2135, EC 1.2.4.1, EC 6.1.1.3, Electroneutral potassium-chloride cotransporter 3, KCC3A, KCC3B, KCC3potassium chloride cotransporter 3, K-Cl cotransporter 3, potassium chloride cotransporter KCC3a-S3, potassium-chloride transporter-3a, potassium-chloride transporter-3b, solute carrier family 12 (potassium/chloride transporters), member 6, solute carrier family 12 member 6
Host Species Rabbit
Immunogen Between 75 and 125
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 9990
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.