missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCC2/SLC12A5 Antibody (905705), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8369MFV450
This item is not returnable.
View return policy
Description
KCC2/SLC12A5 Monoclonal antibody specifically detects KCC2/SLC12A5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| KCC2/SLC12A5 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 905705 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5 | |
| NS0 mouse myeloma cell line transfected with human SLC12A5, Accession # Q9H2X9 | |
| 0.1 mL | |
| Potassium Channels | |
| 57468 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction