missing translation for 'onlineSavingsMsg'
Learn More

KCC2/SLC12A5 Antibody (905705) [Janelia Fluor™ 525], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB8369JF525

Product Code. 30171931

  • £459.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

KCC2/SLC12A5 Monoclonal antibody specifically detects KCC2/SLC12A5 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KCC2/SLC12A5
Monoclonal
Janelia Fluor 525
Electroneutral potassium-chloride cotransporter 2, hKCC2, KCC2K-Cl cotransporter 2, KIAA1176erythroid K-Cl cotransporter 2, Neuronal K-Cl cotransporter, solute carrier family 12 (potassium/chloride transporter), member 5, solute carrier family 12 member 5
NS0 mouse myeloma cell line transfected with human SLC12A5, Accession # Q9H2X9
RUO
Primary
Human
Liquid
Flow Cytometry, Western Blot, CyTOF
905705
50mM Sodium Borate
Mouse
0.1 mL
Potassium Channels
57468
Store at 4°C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.