missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCC2 Recombinant Mouse Monoclonal Antibody (GT16)

KCC2 Recombinant Mouse Monoclonal Antibody (GT16)
Brand: Invitrogen MA556769
This item is not returnable.
View return policy
Description
KCC2 Recombinant Monoclonal Antibody for Western Blot
Specifications
| KCC2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q91V14 | |
| SLC12A5 | |
| The immunogen used to generate this antibody corresponds to human KCC2. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| GT16 | |
| Unconjugated | |
| SLC12A5 | |
| EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5 | |
| Mouse | |
| Protein A | |
| RUO | |
| 57138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction