Learn More
KCC2 Monoclonal Antibody (S1-12), PerCP, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen MA545497
Additional Details : Weight : 0.01000kg
Product Code. 17848402
  • £362.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
1 µg/mL of MA5-45497 was sufficient for detection of KCC2 in 10 µg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 140kDa.
Mediates electroneutral potassium-chloride cotransport in mature neurons. Transport occurs under isotonic conditions, but is activated 20-fold by cell swelling. Important for Cl(-) homeostasis in neurons.
Specifications
Specifications
KCC2
Monoclonal
1 mg/mL
2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4
Q63633, Q91V14, Q9H2X9
SLC12A5
Fusion protein amino acids 932-1043 corresponding to rat KCC2
100 μg
Primary
Human, Mouse, Rat
Liquid
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
S1-12
PerCP
SLC12A5
EIEE34; EIG14; Electroneutral potassium-chloride cotransporter 2; erythroid K-Cl cotransporter 2; furosemide-sensitive K-Cl cotransporter; hKCC2; KCC 2; Kcc2; KCC2a variant 1; KCC2a-S25 variant 1; KCC2b variant 2; K-Cl cotransporter 2; KIAA1176; mKCC2; mKIAA1176; neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; S12A5; SLC12A5; solute carrier family 12 (potassium/chloride transporter), member 5; solute carrier family 12 (potassium-chloride transporter), member 5; solute carrier family 12 member 5; solute carrier family 12, (potassium-chloride transporter) member 5; solute carrier family 12, member 5
Mouse
Protein G
RUO
171373, 57138, 57468
4° C
IgG2a