missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542444
This item is not returnable.
View return policy
Description
Peptide sequence: MPHFTVVPVD GPRRGDYDNL EGLSWVDYGE RAELDDSDGH GNHRESSPFL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%.
SLC12A4 mediates electroneutral potassium-chloride cotransport when activated by cell swelling. SLC12A4 may contribute to cell volume homeostasis in single cells.
Specifications
| KCC1 | |
| Polyclonal | |
| Unconjugated | |
| SLC12A4 | |
| AW546649; CTC-479C5.17; Electroneutral potassium-chloride cotransporter 1; erythroid K-Cl cotransporter 1; FLJ40489; Furosemide-sensitive K-Cl cotransporter; hKCC1; Kcc1; K-Cl cotransporter; K-Cl cotransporter 1; K-Cl cotransporter 4; K-Cl cotransporter KCC1; K-Cl Co-transporter-1; mKCC1; potassium chloride cotransporter SLC12A4; potassium/chloride cotransporter 1; potassium-chloride cotransporter-1; RBCKCC1; RBKCC1; rKCC1; Slc12a4; solute carrier family 12 (potassium/chloride transporter), member 4; solute carrier family 12 (potassium/chloride transporters), member 4; solute carrier family 12 member 4; solute carrier family 12, member 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6560 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UP95 | |
| SLC12A4 | |
| Synthetic peptide directed towards the N-terminal of human SLC12A4 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction