missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KBTBD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35862-100ul
This item is not returnable.
View return policy
Description
KBTBD8 Polyclonal antibody specifically detects KBTBD8 in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| KBTBD8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| kelch repeat and BTB (POZ) domain containing 8, KIAA1842, TAKRP, T-cell activation kelch repeat protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-280 of human KBTBD8 (NP_115894.2).,, Sequence:, DIVVEVDHGKTFSCHRNVLAAISPYFRSMFTSGLTESTQKEVRIVGVEAESMDLVLNYAYTSRVILTEANVQALFTAASIFQIPSIQDQCAKYMISHLDPQNSIGVFIFADHYGHQELGDRSKEYIRKKFLCVTKEQEFLQLTKDQLISILDSDDLNVDREEHVYESIIRWFEHEQNEREVHLPEIFAKCIRFPLMEDTFIEKIPPQFAQAIAKSCVEKGPSNTNGCTQRL | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84541 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction