missing translation for 'onlineSavingsMsg'
Learn More

KBTBD8 Antibody (7G8A6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£173.00 - £435.00

Specifications

Antigen KBTBD8
Clone 7G8A6
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18188619
View Documents
Novus Biologicals
NBP2-52496
0.1 mg
£435.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18696837
View Documents
Novus Biologicals
NBP2-52496-0.025mg
0.025 mg
£173.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KBTBD8 Monoclonal specifically detects KBTBD8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications

Specifications

KBTBD8
1 mg/mL
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry
Unconjugated
Mouse
Human
kelch repeat and BTB (POZ) domain containing 8, KIAA1842, TAKRP, T-cell activation kelch repeat protein
KBTBD8
IgG1
Protein A or G purified
7G8A6
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Monoclonal
Purified
RUO
PBS with 0.05% Sodium Azide
84541
Purified recombinant fragment of human KBTBD8 (AA: 264-464) expressed in E. Coli.
Primary
68.8 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.