missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KBTBD7 Antibody (OTI1G3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72268CL1
This item is not returnable.
View return policy
Description
KBTBD7 Monoclonal antibody specifically detects KBTBD7 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| KBTBD7 | |
| Monoclonal | |
| CoraFluor 1 | |
| DKFZP434E2318, kelch repeat and BTB (POZ) domain containing 7, kelch repeat and BTB domain-containing protein 7 | |
| Full length human recombinant protein of human KBTBD7 (NP_115514) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Biology | |
| 84078 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1G3 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction