missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KBTBD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540948
This item is not returnable.
View return policy
Description
Peptide sequence: LRKVQMVKDA HEGRITTLRK KKKGKDGAGA KEADKGTSKA KAEEDEEAER Sequence homology: Horse: 79%; Human: 100%.
The exact function of KBTBD5 remains unknown.
Specifications
| KBTBD5 | |
| Polyclonal | |
| Unconjugated | |
| Klhl40 | |
| 2310024D23Rik; KBTBD5; kelch like family member 40; kelch repeat and BTB (POZ) domain containing 5; kelch repeat and BTB domain-containing protein 5; kelch-like 40; kelch-like family member 40; kelch-like protein 40; kelch-like protein 40; LOW QUALITY PROTEIN: kelch-like protein 40; Klhl40; NEM8; nemaline myopathy type 8; sarcosynapsin; SRYP; SYRP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 131377 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q2TBA0 | |
| Klhl40 | |
| Synthetic peptide directed towards the middle region of human KBTBD5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction