missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KBTBD10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25577-1-AP-150UL
This item is not returnable.
View return policy
Description
KLHL41 is involved in skeletal muscle development and differentiation. It regulates proliferation and differentiation of myoblasts and plays a role in myofibril assembly by promoting lateral fusion of adjacent thin fibrils into mature, wide myofibrils. It is required for pseudopod elongation in transformed cells.Specifications
| KBTBD10 | |
| Polyclonal | |
| Unconjugated | |
| KLHL41 | |
| KBTBD10, Kel like protein 23, Kelch related protein 1, KRP1, SARCOSIN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10324, 228003 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A2AUC9, O60662 | |
| KLHL41 | |
| KBTBD10 Fusion Protein Ag22311 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction