Learn More
Specac™ KBr Sealed Heatable Liquid Flow Cell VTCH Incompatible
Product Code. 15210228
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Type:
KBr Sealed Heatable Liquid Flow Cell
Product Code. Type
15210228 KBr Sealed Heatable Liquid Flow Cell
1 missing translation for 'options'
Product Code. 15210228 missing translation for 'mfr' Specac™ missing translation for 'supplierNo' GS20571
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
This cell is designed for use in Specac heating jackets GS20710 and GS20730. This is a sealed heatable liquid flow cell with KBr windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Type KBr Sealed Heatable Liquid Flow Cell
Description Pathlength from 0.025 mm up to 1.0 mm