Learn More
Specac™ KBr Demountable Heatable Liquid Flow Cell VTCH Incompatible
Product Code. 15260228
Encompass_Preferred
Change view
Click to view available options
Type:
KBr Demountable Heatable Liquid Flow Cell
Product Code. Type
15260228 KBr Demountable Heatable Liquid Flow Cell
1 options
Product Code. 15260228 Supplier Specac™ Supplier No. GS20581
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This cell is designed for use in Specac heating jackets GS20710 and GS20730. This is a demountable heatable liquid flow cell with KBr windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Type KBr Demountable Heatable Liquid Flow Cell
Description Pathlength from 0.025 mm up to 1.0 mm