missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KBP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA557313

Product Code. 15745211

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EIAHAYYDMM DLKVAIADRL RDPDSHIVKK INNLNKSALK YYQLFLDSLR DPNKVFPEHI GEDVLRPAML AKFRVARLYG KIITADPKK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 94%.

Required for neuronal development and differentiation. Required for organization of axonal microtubules, and axonal outgrowth and maintenance during peripheral and central nervous system development. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KBP
Polyclonal
Unconjugated
KIFBP
0710007C18Rik; 2510003E04Rik; hypothetical protein LOC527433; hypothetical protein LOC606294; Kbp; Kiaa1279; KIF1 binding protein; KIF1-binding protein; KIF1-binding protein-like protein; Kif1bp; KIF-binding protein; KIFBP; kinesin binding protein; kinesin family binding protein; LOC606294; mKIAA1279; TTC20
Rabbit
Antigen Affinity Chromatography
RUO
26128
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96EK5
KIFBP
Recombinant protein corresponding to Human KBP. Recombinant protein control fragment (Product #RP-97218).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.