missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KBP Monoclonal Antibody (K06355_16B10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556059
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: DTPEAEGEVP ELYHQRKGEI ARCWIKYCLT LMQNAQLSMQ DNIGELDLDK QSELRALRKK ELDEEESIRK KAVQFGTGEL CDAISAVEEK VSYLRPLDFE EARELFLLGQ HYVFEAKEFF QIDGYVTDHI
Required for neuronal development and differentiation. Required for organization of axonal microtubules, and axonal outgrowth and maintenance during peripheral and central nervous system development. [UniProt].
Specifications
| KBP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q6ZPU9, Q96EK5 | |
| KIF1BP, KIFBP | |
| Recombinant human KIFBP (immunogen range is 291-420). | |
| 200 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| K06355_16B10 | |
| Unconjugated | |
| KIFBP | |
| 0710007C18Rik; 2510003E04Rik; hypothetical protein LOC527433; hypothetical protein LOC606294; Kbp; Kiaa1279; KIF1 binding protein; KIF1-binding protein; KIF1-binding protein-like protein; Kif1bp; KIF-binding protein; KIFBP; kinesin binding protein; kinesin family binding protein; LOC606294; mKIAA1279; TTC20 | |
| Mouse | |
| Protein A | |
| RUO | |
| 26128, 72320 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction