missing translation for 'onlineSavingsMsg'
Learn More

KB-R7943 Mesylate, TRC

Product Code. 30395282
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395282 2.5 mg 2.5mg
30394010 1 mg -
30397152 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395282 Supplier Toronto Research Chemicals Supplier No. TRCK4754002.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182004-65-5
Molecular Formula C16H17N3O3S . (CH4O3S)
Molecular Weight (g/mol) 427.5
Synonym KB-R7943 Mesylate, 2-[4-[(4-Nitrophenyl)methoxy]phenyl]ethyl Ester, Monomethanesulfonate Carbamimidothioic Acid (9CI), 2-[4-[(4-Nitrophenyl)methoxy]phenyl]ethyl Carbamimidothioate Methanesulfonate (1:1), KB-R 7943
IUPAC Name methanesulfonic acid;2-[4-[(4-nitrophenyl)methoxy]phenyl]ethyl carbamimidothioate
SMILES CS(=O)(=O)O.NC(=N)SCCc1ccc(OCc2ccc(cc2)[N+](=O)[O-])cc1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C16H17N3O3S.CH4O3S/c17-16(18)23-10-9-12-3-7-15(8-4-12)22-11-13-1-5-14(6-2-13)19(20)21;1-5(2,3)4/h1-8H,9-11H2,(H3,17,18);1H3,(H,2,3,4)
Formula Weight 427.0872
Chemical Name or Material 2-[2-[4-(4-Nitrobenzyloxy)phenyl]ethyl]isothiourea Mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.