missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KAZALD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580749
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KAZALD1.
This gene encodes a secreted member of the insulin growth factor-binding protein (IGFBP) superfamily. It contains an N-terminal insulin growth factor-binding domain, a central Kazal-type serine protease inhibitor and follistatin-like domain, and a C-terminal immunoglobulin-like domain. Studies of the mouse ortholog suggest that this gene product may have a function in bone development and bone regeneration.
Specifications
| KAZALD1 | |
| Polyclonal | |
| Unconjugated | |
| KAZALD1 | |
| bone- and odontoblast-expressed gene 1; BONO1; FKSG28; FKSG40; FLJ24094; IGFBP-related protein 10; IGFBP-rP10; Kazal type serine peptidase inhibitor domain 1; KAZALD1; kazal-type serine protease inhibitor domain-containing protein 1; novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein; UNQ2945/PRO21184 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 81621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q96I82 | |
| KAZALD1 | |
| A synthetic peptide corresponding to the center region of the Human KAZALD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction