missing translation for 'onlineSavingsMsg'
Learn More

Kawain, TRC

Product Code. 30395034
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395034 200 mg 200mg
30406224 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395034 Supplier Toronto Research Chemicals Supplier No. TRCK145700200MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-64-1
Molecular Formula C14 H14 O3
Molecular Weight (g/mol) 230.26
Synonym [R-(E)]-5,6-Dihydro-4-methoxy-6-(2-phenylethenyl)-2H-pyran-2-one, (R)-5,6-Dihydro-4-methoxy-6-styryl-2H-pyran-2-one, (+)-Kavain, (+)-Kawain, (R)-(+)-Kavain, Gonosan, Kavain, Kawain, NSC 112162, d-Kawain
IUPAC Name (2R)-4-methoxy-2-[(E)-2-phenylethenyl]-2,3-dihydropyran-6-one
SMILES COC1=CC(=O)O[C@H](C1)\C=C\c2ccccc2

Specifications

Quantity 200 mg
InChI Formula InChI=1S/C14H14O3/c1-16-13-9-12(17-14(15)10-13)8-7-11-5-3-2-4-6-11/h2-8,10,12H,9H2,1H3/b8-7+/t12-/m0/s1
Formula Weight 230.0943
Chemical Name or Material Kawain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.