missing translation for 'onlineSavingsMsg'
Learn More

Nordvlies Katrin Basic Toilet 250

Product Code. 15730937
Change view
Click to view available options
Color:
White
Unit Size:
64 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15730937 White 64 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15730937 Supplier Nordvlies Supplier No. 16950
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Basic Toilet 250 is conventional toilet paper for everyday use.

  • Conventional toilet paper for everyday use
  • 2-Ply, natural white
  • Lenght: 250 sheets, 29 m
  • Good, absorbent paper for maximum comfort
  • Certified under the Nordic Swan Eco Label
  • All Katrin toilet papers are made from high quality fibers and have been proven, via Dissolving test 90 seconds, Metsä Tissue 02 method, to break down and dissolve quickly and efficiently in water, under normal usage conditions
  • Certified under the EU Flower Eco Label

Specifications

Product Type Toilet Paper
Length (Metric) 29 m
Certifications/Compliance Nordic Swan Eco Label
Color White
For Use With (Equipment) Katrin System Toilet Dispenser
Packaging Quantity 250 Sheets
Ply 2-ply
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.