missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KATNAL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113096
This item is not returnable.
View return policy
Description
KATNAL2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| KATNAL2 | |
| Polyclonal | |
| Unconjugated | |
| KATNAL2 | |
| katanin catalytic subunit A like 2; katanin catalytic subunit A1 like 2; katanin p60 ATPase-containing subunit A-like 2; katanin p60 subunit A like 2; Katanin p60 subunit A-like 2; KATNAL2; p60 katanin-like 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83473 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IYT4 | |
| KATNAL2 | |
| Recombinant Human Katanin p60 ATPase-containing subunit A-like 2 protein (18-307AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction