missing translation for 'onlineSavingsMsg'
Learn More

Katanin p60 Antibody [DyLight 755], Novus Biologicals™

Product Code. 30735603 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735603 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735603 Supplier Novus Biologicals Supplier No. AF7100Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Katanin p60 Polyclonal antibody specifically detects Katanin p60 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Katanin p60
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias EC 3.6.4.3, katanin p60 (ATPase containing) subunit A 1, katanin p60 (ATPase-containing) subunit A 1, katanin p60 ATPase-containing subunit A1, Katanin p60 subunit A1, p60 katanin
Host Species Sheep
Immunogen E. coli-derived recombinant human Katanin p60, Asp308-Tyr378, Accession # O75449
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 11104
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.