missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT6B-MORF Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
KAT6B-MORF Polyclonal specifically detects KAT6B-MORF in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | KAT6B-MORF |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | DKFZp313G1618, FLJ90335, Histone acetyltransferase MORF, Histone acetyltransferase MOZ2, histone acetyltransferase MYST4, KAT6B, KIAA0383, Monocytic leukemia zinc finger protein-related factor, MORFMorf, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 4, MOZ2EC 2.3.1.48, MOZ-related factor, MYST histone acetyltransferase (monocytic leukemia) 4, MYST-4, qkf, querkopf |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 400-500 of human KAT6B-MORF (NP_001243398.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?