missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | KAT5 |
|---|---|
| Concentration | 0.43 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817473
|
Proteintech
10827-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807473
|
Proteintech
10827-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIP60 is a transcription factor, originally isolated as an HIV-1 TAT-interactive protein, known to interact with the viral TAT protein that leads to HTATIP polyubiquitination and targets it for degradation. It belongs to the MYST family of histone acetyl transferases (HATs). TIP60 is a catalytic subunit of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. A co-regulator of transcription factors, TIP60 either promotes or suppresses tumorigenesis. TIP60 is an enzyme that has a role in DNA repair and apoptosis and is thought to play an important role in signal transduction.Specifications
| KAT5 | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q8CHK4, Q92993, Q99MK2 | |
| 10524, 192218, 81601 | |
| KAT5 Fusion Protein Ag1261 | |
| Primary | |
| -20°C | |
| KAT5 |
| 0.43 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 60 kDa Tat interactive protein, cPLA(2) interacting protein, cPLA2, ESA1, Histone acetyltransferase KAT5, HIV 1 Tat interactive protein, HTATIP, HTATIP1, K(lysine) acetyltransferase 5, KAT5, Lysine acetyltransferase 5, PLIP, TIP, TIP60 | |
| KAT5 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title