missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT5 Mouse anti-Human, Clone:4B3B4, Abnova™
Mouse monoclonal antibody raised against recombinant human KAT5.
Brand: Abnova MAB16675.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the MYST family of histone acetyl transferases (HATs) and was originally isolated as an HIV-1 TAT-interactive protein. HATs play important roles in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and nonhistone proteins. This protein is a histone acetylase that has a role in DNA repair and apoptosis and is thought to play an important role in signal transduction. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]Specifications
| KAT5 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry The optimal working dilution should be determined by the end user. |
|
| KAT5 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 4B3B4 | |
| K(lysine) acetyltransferase 5 | |
| In PBS (0.05% sodium azide) | |
| ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, cPLA2 | |
| Recombinant protein corresponding to amino acid 18-208 of human KAT5 from E. coli. | |
| 10524 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction