missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT4/TBP Associated Factor 1 Antibody (SR1493), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22390-50ul
This item is not returnable.
View return policy
Description
KAT4/TBP Associated Factor 1 Monoclonal antibody specifically detects KAT4/TBP Associated Factor 1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| KAT4/TBP Associated Factor 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1493 | |
| Western Blot 1:500-1:2000 | |
| 250kDa, BA2Rdystonia 3 (with Parkinsonism), CCG1TATA box binding protein (TBP)-associated factor, RNA polymerase II, A, 250kD, CCGS, Cell cycle gene 1 protein, cell cycle, G1 phase defect, complementation of cell cycle block, G1-to-S, DYT3/TAF1, EC 2.7.11, EC 2.7.11.1, KAT4, NSCL2, OF, P250, TAF(II)250, TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2AN-TAF1, TAFII-250, TAFII250DYT3, TBP-associated factor 250 kDa, transcription factor TFIID p250 polypeptide, Transcription initiation factor TFIID 250 kDa subunit, transcription initiation factor TFIID subunit 1, XDP | |
| A synthesized peptide derived from human KAT4/TBP Associated Factor 1 (Uniprot #: P21675) | |
| 50 μg | |
| Protein Kinase | |
| 6872 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction