missing translation for 'onlineSavingsMsg'
Learn More

KAT2A/GCN5 Antibody (4F9), Novus Biologicals™

Product Code. 18350649 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350649 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350649 Supplier Novus Biologicals Supplier No. H00002648M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KAT2A/GCN5 Monoclonal antibody specifically detects KAT2A/GCN5 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAT2A/GCN5
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4F9
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH32743
Gene Alias EC 2.3.1.48, GCN5 general control of amino-acid synthesis 5-like 2 (yeast), GCN5L2GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2, GCN5MGC102791, General control of amino acid synthesis protein 5-like 2, General control of amino acid synthesis, yeast, homolog-like 2, HGCN5, Histone acetyltransferase GCN5, histone acetyltransferase KAT2A, HsGCN5, K(lysine) acetyltransferase 2A, Lysine acetyltransferase 2A, PCAF-b, STAF97
Host Species Mouse
Immunogen GCN5L2 (AAH32743, 738 a.a. ~ 837 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KNLLAQIKSHPSAWPFMEPVKKSEAPDYYEVIRFPIDLKTMTERLRSRYYVTRKLFVADLQRVIANCREYNPPDSEYCRCASALEKFFYFKLKEGGLIDK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Epigenetics, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2648
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.