Learn More
KAT2A/GCN5 Antibody (3G13), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-48593MFV450
This item is not returnable.
View return policy
Description
KAT2A/GCN5 Monoclonal antibody specifically detects KAT2A/GCN5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Immunocytochemistry | |
| Monoclonal | |
| Protein G purified | |
| RUO |