missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT2A/GCN5 Antibody (3G13), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-48593FR
This item is not returnable.
View return policy
Description
KAT2A/GCN5 Monoclonal specifically detects KAT2A/GCN5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| KAT2A/GCN5 | |
| Monoclonal | |
| DyLight 680 | |
| Q92830 | |
| KAT2A | |
| Recombinant human GCN5L2 (411-837aa) purified from E. coli (NP_066564). | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3G13 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| EC 2.3.1.48, GCN5 general control of amino-acid synthesis 5-like 2 (yeast), GCN5L2GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2, GCN5MGC102791, General control of amino acid synthesis protein 5-like 2, General control of amino acid synthesis, yeast, homolog-like 2, HGCN5, Histone acetyltransferase GCN5, histone acetyltransferase KAT2A, HsGCN5, K(lysine) acetyltransferase 2A, Lysine acetyltransferase 2A, PCAF-b, STAF97 | |
| Mouse | |
| 94 kDa | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Epigenetics, Signal Transduction | |
| 2648 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction