missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT2A/GCN5 Antibody (3G13), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-48593CL1
This item is not returnable.
View return policy
Description
KAT2A/GCN5 Monoclonal antibody specifically detects KAT2A/GCN5 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| KAT2A/GCN5 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.3.1.48, GCN5 general control of amino-acid synthesis 5-like 2 (yeast), GCN5L2GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2, GCN5MGC102791, General control of amino acid synthesis protein 5-like 2, General control of amino acid synthesis, yeast, homolog-like 2, HGCN5, Histone acetyltransferase GCN5, histone acetyltransferase KAT2A, HsGCN5, K(lysine) acetyltransferase 2A, Lysine acetyltransferase 2A, PCAF-b, STAF97 | |
| Recombinant human KAT2A/GCN5 (411-837aa) purified from E. coli (NP_066564). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Epigenetics, Signal Transduction | |
| 2648 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 3G13 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction