missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT1 Rabbit anti-Human, Mouse, Rat, Clone: 7U2O3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16412-100UL
This item is not returnable.
View return policy
Description
KAT1 Monoclonal antibody specifically detects KAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| KAT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7U2O3 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 2.3.1.48, histone acetyltransferase 1, histone acetyltransferase type B catalytic subunit, KAT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human KAT1 (O14929). | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8520 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction