missing translation for 'onlineSavingsMsg'
Learn More

Kasugamycin hydrochloride hydrate, MedChemExpress

Product Code. 16472359
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 16472359

Brand: Medchem Express HYB1864B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kasugamycin hydrochloride hydrate (Ksg hydrochloride hydrate) is an antibiotic which binds both the 30S and 70S ribosome but not isolated 50S subunits. Kasugamycin hydrochloride hydrate (Ksg hydrochloride hydrate) mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.

Kasugamycin hydrochloride hydrate (Ksg hydrochloride hydrate) is an antibiotic which binds both the 30S and 70S ribosome but not isolated 50S subunits. Kasugamycin hydrochloride hydrate (Ksg hydrochloride hydrate) mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.

Specifications

Chemical Name or Material Kasugamycin hydrochloride hydrate
Physical Form Solid
CAS 200132-83-8
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C14H28ClN3O10
For Use With (Application) COVID-19-immunoregulation
Synonym Ksg hydrochloride hydrate
Solubility Information H2O : 25 mg/mL (57.62 mM; Need ultrasonic) ∣DMSO : 1.67 mg/mL (3.85 mM; ultrasonic and warming and heat to 80°C)
SMILES OC(C(N[C@@H](C[C@@H]1N)[C@@H](C)O[C@]1([H])O[C@@H]2[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)[C@H]2O)=N)=O.[H]Cl.O
Molecular Weight (g/mol) 433.84
Formula Weight 433.84
Percent Purity 99.95%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.