missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karyopherin (importin) beta 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | Karyopherin (importin) beta 3 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673926
|
Novus Biologicals
NBP2-38795-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18163228
|
Novus Biologicals
NBP2-38795 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Karyopherin (importin) beta 3 Polyclonal specifically detects Karyopherin (importin) beta 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Karyopherin (importin) beta 3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp686O1576, FLJ43041, IMB3, imp5, importin 5, importin beta-3 subunit, Importin subunit beta-3, importin-5, karyopherin (importin) beta 3, Karyopherin beta-3, KPNB3, MGC2068, RAN binding protein 5, Ran_GTP binding protein 5, Ran-binding protein 5, RANBP5 | |
| IPO5 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O00410 | |
| 3843 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MPEDQVGKLEATENTISAMA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title