missing translation for 'onlineSavingsMsg'
Learn More

Karyopherin (importin) beta 3 Antibody (1C4), Novus Biologicals™

Product Code. 18396698 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396698 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396698 Supplier Novus Biologicals Supplier No. H00003843M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Karyopherin (importin) beta 3 Monoclonal antibody specifically detects Karyopherin (importin) beta 3 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Karyopherin (importin) beta 3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1C4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry 1:10 to 1:500, Immunohistochemistry-Paraffin 1:10 to 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01497
Gene Alias DKFZp686O1576, FLJ43041, IMB3, imp5, importin 5, importin beta-3 subunit, Importin subunit beta-3, importin-5, karyopherin (importin) beta 3, Karyopherin beta-3, KPNB3, MGC2068, RAN binding protein 5, Ran_GTP binding protein 5, Ran-binding protein 5, RANBP5
Host Species Mouse
Immunogen RANBP5 (AAH01497, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAAAAEQQQFYLLLGNLLSPDNVVRKQAEETYENIPGQSKITFLLQAIRNTTAAEEARQMAAVLLRRLLSSAFDEVYPALPSDVQTAIKSELLMIIQME
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3843
Target Species Human, Mouse, Rat, Canine
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.