missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Kartogenin

Product Code. 18085732
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085732 10 mg 10mg
18084212 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085732 Supplier Tocris Bioscience™ Supplier No. 4513/10
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potently induces chondrogenesis in MSCs

Kartogenin potently induces differentiation of human mesenchymal stem cells into chondrocytes (EC50 = 100 nM). Reduces disease severity in a mouse model of osteoarthritis; displays protective effects against osteoarthritic stimuli in mature chondrocytes in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4727-31-5
Molecular Formula C20H15NO3

Specifications

Quantity 10 mg
Formula Weight 317.34
Purity >98%
Molecular Weight (g/mol) 317.344
Chemical Name or Material Kartogenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.