missing translation for 'onlineSavingsMsg'
Learn More

Kartogenin, MedChemExpress

Product Code. 16499557
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499557 5 mg 5mg
16457076 100 mg 100mg
16453067 50 mg 50mg
16447018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499557 Supplier Medchem Express Supplier No. HY16268
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Kartogenin (KGN) is an inducer of differentiation of human mesenchymal stem cells into chondrocytes, with an EC50 of 100 nM. Kartogenin binds filamin A, disrupts its interaction with the transcription factor core-binding factor β subunit (CBFβ), and induces chondrogenesis by regulating the CBFβ-RUNX1 transcriptional program. Kartogenin can be used for the research of osteoarthritis (OA).

Kartogenin (KGN) is an inducer of differentiation of human mesenchymal stem cells into chondrocytes, with an EC50 of 100 nM. Kartogenin binds filamin A, disrupts its interaction with the transcription factor core-binding factor β subunit (CBFβ), and induces chondrogenesis by regulating the CBFβ-RUNX1 transcriptional program. Kartogenin can be used for the research of osteoarthritis (OA).

Specifications

Chemical Name or Material Kartogenin
Physical Form Powder
CAS 4727-31-5
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H15NO3
Synonym KGN
Solubility Information DMSO : ≥ 42 mg/mL (132.35 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(O)C1=CC=CC=C1C(NC2=CC=C(C3=CC=CC=C3)C=C2)=O
Molecular Weight (g/mol) 317.34
Formula Weight 317.34
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.